Methanol ≥99,8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Collection
Gefahr

20847.990EA 0 EUR
20847.990
Methanol ≥99,8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Methanol
Formel: CH₃OH
Siedepunkt: 64,6 °C (1013 hPa)
Schmelzpunkt: –98 °C
Dichte: 0,7918 g/cm³ (20 °C)
Flammpunkt: 11 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004595
CAS-Nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Clear colourless liquid
Assay (on anhydrous substance) Min. 99.8 %
IR Spectrum Passes test
Solubility in water Passes test ACS
Acidity Max. 0.0002 meq/g
Alkalinity Max. 0.00006 meq/g
Boiling point 64 - 65 °C
Colouration Max. 10 APHA
Density (20/4) 0.791 - 0.792
Density (20/20) 0.791 - 0.793
Substances coloured by H2SO4 Max. 10 APHA
Acetone + aldehydes (as CH3COCH3) Max. 10 ppm
Acetone Max. 5 ppm
Acetaldehyde Max. 5 ppm
Ethanol Max. 0.1 %
Evaporation residue Max. 10 ppm
Formaldehyde Max. 1 ppm
Substances reducing KMnO4 (as O) Max. 2 ppm
Water Max. 0.05 %
Cl (Chloride) Max. 0.5 ppm
Al (Aluminium) Max. 0.1 ppm
B (Boron) Max. 0.05 ppm
Ba (Barium) Max. 0.05 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 0.7 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.2 ppm
Absorbance (225 nm) Max. 0.17
Transmittance (210 nm) Min. 20 %
Transmittance (220 nm) Min. 50 %
Transmittance (225 nm) Min. 68 %
Transmittance (250 nm) Min. 95 %
Conforms to ACS Passes test
Conforms Ph.Eur. R 1053200 Passes test
Conforms Ph.Eur. R1 1053201 Passes test
Conforms Ph.Eur. R2 1053202 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR