1-Propanol ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

Synonyme: 1-Propylalkohol, 1-Hydroxypropan, Ethylcarbinol

20861.294EA 85.8 EUR
20861.294 20861.363 20861.465 20861.320 20861.550
1-Propanol ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
1-Propanol
Formel: H₃CCH₂CH₂OH
Molecular Weight: 60.1 g/mol
Siedepunkt: 97
Schmelzpunkt: –127 °C
Dichte: 0.8053 g/cm³ (20 °C. DIN 51757)
Flammpunkt: 15
Lagertemperatur: Raumtemperatur
MDL: MFCD00002941
CAS-Nummer: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (on anhydrous substance) Min. 99.5 %
Distillation range Passes test Reag.Ph.Eur.
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0006 meq/g
Boiling point 96.9 - 97.5 °C
Colouration Max. 10 APHA
Density (20/4) 0.801 - 0.804
Density (20/20) 0.802 - 0.806
Acetone Max. 100 ppm
Evaporation residue Max. 10 ppm
Ethanol Max. 0.05 %
Methanol Max. 0.05 %
Propan-2-ol Max. 0.05 %
Propionaldehyde Max. 0.06 %
Water Max. 0.1 %
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.05 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR