1-Propanol ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
1-Propylalkohol, 1-Hydroxypropan, Ethylcarbinol
20861.294EA
85.8
EUR
20861.294
20861.363
20861.465
20861.320
20861.550
1-Propanol ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
1-Propanol
Formel:
H₃CCH₂CH₂OH Molecular Weight: 60.1 g/mol Siedepunkt: 97 Schmelzpunkt: –127 °C Dichte: 0.8053 g/cm³ (20 °C. DIN 51757) Flammpunkt: 15 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002941 CAS-Nummer: 71-23-8 EINECS: 200-746-9 UN: 1274 ADR: 3 II Merck Index: 13,07934 |
|
Spezifikation Testergebnisse
Assay (on anhydrous substance) | Min. 99.5 % |
Distillation range | Passes test Reag.Ph.Eur. |
Acidity | Max. 0.0005 meq/g |
Alkalinity | Max. 0.0006 meq/g |
Boiling point | 96.9 - 97.5 °C |
Colouration | Max. 10 APHA |
Density (20/4) | 0.801 - 0.804 |
Density (20/20) | 0.802 - 0.806 |
Acetone | Max. 100 ppm |
Evaporation residue | Max. 10 ppm |
Ethanol | Max. 0.05 % |
Methanol | Max. 0.05 % |
Propan-2-ol | Max. 0.05 % |
Propionaldehyde | Max. 0.06 % |
Water | Max. 0.1 % |
Al (Aluminium) | Max. 0.5 ppm |
B (Boron) | Max. 0.02 ppm |
Ba (Barium) | Max. 0.1 ppm |
Ca (Calcium) | Max. 0.5 ppm |
Cd (Cadmium) | Max. 0.05 ppm |
Co (Cobalt) | Max. 0.02 ppm |
Cr (Chromium) | Max. 0.02 ppm |
Cu (Copper) | Max. 0.05 ppm |
Fe (Iron) | Max. 0.1 ppm |
Mg (Magnesium) | Max. 0.1 ppm |
Mn (Manganese) | Max. 0.02 ppm |
Ni (Nickel) | Max. 0.02 ppm |
Pb (Lead) | Max. 0.1 ppm |
Sn (Tin) | Max. 0.1 ppm |
Zn (Zinc) | Max. 0.1 ppm |
Conforms to Reag. Ph.Eur. | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...