Cadmiumacetat Dihydrat ≥98,0% (durch komplexometrische Analyse), Purum p.a.

Lieferant: Honeywell Chemicals
Gefahr

20901-500G
20901-500GEA 128.06 EUR
20901-500G
Cadmiumacetat Dihydrat ≥98,0% (durch komplexometrische Analyse), Purum p.a.
Cadmiumacetat Dihydrat

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formel: (H₃CCOO)₂Cd·2H₂O
Siedepunkt: Zersetzung
Schmelzpunkt: 256 °C,anhydrous substance.Elimination of water of crystallisation
Dichte: 2,01 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150020
CAS-Nummer: 5743-04-4
EINECS: 208-853-2
ADR: 6.1,III
Merck Index: 13,01614

Bestellen

Spezifikation Testergebnisse

Assay 98.0 - 102.0 %
Calcium (Ca) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Sodium (Na) Max. 100 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 50 ppm
Sulfate (SO₄) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR