Tetramethylammoniumhydroxid 25% w/w aqueous solution , 99,9999% Metals basis, Electronic Grade for the electronics industry
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
20932.AK
20932.K2
20932.AKEA
53.8
EUR
20932.AK
20932.K2
Tetramethylammoniumhydroxid 25% w/w aqueous solution , 99,9999% Metals basis, Electronic Grade for the electronics industry
Tetramethylammonium hydroxide
Purity: 99.9999%
CAS No.: 75-59-2
Molecular Formula: C4H13NO
Formula Weight: 91.15
Physical Form: Liquid
Density: 1.016
Refractive Index: 1.3840
Sensitivity: Air Sensitive
Appearance: Colourless to pale yellow
Boiling Point: ca 102°
Beilstein Reference: 3558708
Merck Reference: 14,9224
UN No.: 1835
MDL No.: MFCD00008280
Formel:
C₄H₁₃NO Molecular Weight: 91.15 g/mol Siedepunkt: ∼102 °C Dichte: 1.016 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008280 CAS-Nummer: 75-59-2 EINECS: 200-882-9 UN: 1835 ADR: 8 II Merck Index: 14,9224 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...