Devardas Legierung, Granulat, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals

20942.181EA 143 EUR
20942.181 20942.294
Devardas Legierung, Granulat, AnalaR NORMAPUR® analytisches Reagens
Devardas Legierung

Devarda's alloy is used as reducing agent in analytical chemistry for the determination of nitrates after their reduction to ammonia under alkaline.


  • For reduction reagent for total Nitrogen determination acc. Kjeldahl
  • For reduction of alkaline Chlorate solutions to Chloride


Devarda's alloy (CAS # 8049-11-4) is an alloy of aluminium (44% – 46%), copper (49% – 51%) and zinc (4% – 6%).

Siedepunkt: 906 °C (1013 hPa)
Schmelzpunkt: 490…560 °C
Dichte: 5,79 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00198176
CAS-Nummer: 8049-11-4
UN: 3089
ADR: 4.1,II
Merck Index: 13,02958

Bestellen

Spezifikation Testergebnisse

N (Stickstoff) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR