Aluminiumoxid, Durmax® für die Metallographie, 3 Stunden
Lieferant: ALUMINES DURMAX
Synonyme:
Aluminium(III) oxide, Alumina C, Aluminum oxide
20992.263
20992.263EA
84.8
EUR
20992.263
Aluminiumoxid, Durmax® für die Metallographie, 3 Stunden
Aluminium oxide
Lieferumfang: This product is not available in all countries. Please check with your local VWR International office or supplier.
Formel:
Al₂O₃ Molecular Weight: 101.96 g/mol Siedepunkt: 2980 Schmelzpunkt: 2045 Dichte: 3.94 g/cm³ (20 °C) |
MDL:
MFCD00003424 CAS-Nummer: 1344-28-1 EINECS: 215-691-6 REACH: 01-2119529248-35 Merck Index: 13,00355 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...