Ethylmagnesiumbromid 0.9 M in THF, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
210471000
210478000
210471000.EA
57.1
EUR
210471000.
210478000.
Ethylmagnesiumbromid 0.9 M in THF, AcroSeal®
Ethylmagnesium bromide
Formel:
C₂H₅BrMg Flammpunkt: –17 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000043 CAS-Nummer: 925-90-6 EINECS: 213-127-3 UN: 3399 ADR: 4.3,I |
Spezifikation Testergebnisse
Appearance | Brown to grey-brown to brown-black Solution (May develop some turbidity or precipitate) |
Titration Grignard | 0.85 to 1.15 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...