Ethylmagnesiumbromid 0.9 M in THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

210471000 210478000
210471000.EA 57.1 EUR
210471000. 210478000.
Ethylmagnesiumbromid 0.9 M in THF, AcroSeal®
Ethylmagnesium bromide
Formel: C₂H₅BrMg
Flammpunkt: –17 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000043
CAS-Nummer: 925-90-6
EINECS: 213-127-3
UN: 3399
ADR: 4.3,I

Bestellen

Spezifikation Testergebnisse

Appearance Brown to grey-brown to brown-black Solution (May develop some turbidity or precipitate)
Titration Grignard 0.85 to 1.15 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR