Aluminiumsulfat Tetradecahydrat, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Gefahr
|
21070.297EA
71.1
EUR
21070.297
Aluminiumsulfat Tetradecahydrat, GPR RECTAPUR®
Aluminiumsulfat Tetradecahydrat
Formel:
Al₂(SO₄)₃·14H₂O |
CAS-Nummer:
16828-12-9 EINECS: 233-135-0 |
Spezifikation Testergebnisse
Assay (calculated as Al2O3) | 16.5 - 17.5 % |
Heavy metals (as Pb) | Max. 20 ppm |
Not precipitated by NH4OH (as SO4) | Max. 0.3 % |
Cl (Chloride) | Max. 100 ppm |
Fe (Iron) | Max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...