VWR® Pop-Up™, Gestelle für Zentrifugenröhrchen, zusammenklappbar

Lieferant: VWR Collection
211-0368EA 56.5 EUR
211-0368 211-0367 525-0510 525-0511
VWR® Pop-Up™, Gestelle für Zentrifugenröhrchen, zusammenklappbar
Gestelle Reagenzglasgestelle
PP

  • Simple easy action to open and close
  • Designed to help organise storage of 15 and 50 ml tubes
  • Stackable in open or closed position
  • Imprinted grid aids quick identification and retrieval
  • Collapse flat (height 21 mm) for compact storage

Close flat for storage and 'pop up' for quick use.

Note: Robust PP construction but not recommended for cleaning in an autoclave

Bitte beachten Widerstandsfähige PP-Konstruktion, jedoch nicht für die Reinigung in Autoklaven geeignet
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR