Microtubes with cap, 1,5 ml
Lieferant: SEMADENI
1604
211-2081EA 61.1 EUR
211-2081
Microtubes with cap, 1,5 ml
Röhrchen Reaktionsgefäße
PP, translucent.
  • Pierceable lid
  • Autoclavable
  • Withstand centrifugation at 21000×g
  • Excellent chemical resistance
With attached push-on lid.