Aluminiumkaliumsulfat Dodecahydrat 99,5-100,5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Lieferant: VWR Chemicals
Synonyme:
Alumen, Kalialaun, Aluminiumkaliumbis(sulfat) Dodecahydrat
21110.296EA
125
EUR
21110.296
Aluminiumkaliumsulfat Dodecahydrat 99,5-100,5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Aluminiumkaliumsulfat Dodecahydrat
Formel:
AlK(SO₄)₂·12H₂O Molecular Weight: 474,39 g/mol Schmelzpunkt: 92,5 °C Dichte: 1,72 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149143 CAS-Nummer: 7784-24-9 EINECS: 233-141-3 REACH: 01-2119960162-44 Merck Index: 12,00373 |
Spezifikation Testergebnisse
Assay | 99.5 - 100.5 % |
Appearance of solution | Passes test Ph.Eur. |
Identification A | Passes test Ph.Eur. |
Identification B | Passes test Ph.Eur. |
Identification C | Passes test Ph.Eur. |
pH (20°C; 10 %) | 3.0 - 3.5 |
Heavy metals (as Pb) | Max. 10 ppm |
Insolubility in water | Max. 50 ppm |
Cl (Chloride) | Max. 5 ppm |
NH4 (Ammonium) | Max. 50 ppm |
Fe (Iron) | Max. 5 ppm |
Na (Sodium) | Max. 0.02 % |
Pb (Lead) | Max. 5 ppm |
Conforms to ACS | Passes test |
Conforms to Reag. Ph.Eur. | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...