Cerium(III) acetylacetonate hydrate
Lieferant: Thermo Fisher Scientific
|
Achtung
|
21116.22
21116.36
21116.22EA
261
EUR
21116.22
21116.36
Cerium(III) acetylacetonate hydrate
Cerium(III) acetylacetonate hydrate
CAS No.: 206996-61-4
Molecular Formula: C15H21CeO6.xH2O
Formula Weight: 437.44(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Yellow to red
Melting Point: 131-132°
MDL No.: MFCD00150164
Formel:
C₁₅H₂₁CeO₆·xH₂O Schmelzpunkt: 131…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150164 CAS-Nummer: 206996-61-4 EINECS: 239-720-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...