Certrifluorid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
21118.14
21118.22
21118.36
21118.14EA
89.1
EUR
21118.14
21118.22
21118.36
Certrifluorid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Certrifluorid
Purity: 99.9%
CAS No.: 7758-88-5
Molecular Formula: CeF3
Formula Weight: 197.12
Physical Form: Powder
Density: 6.16
Melting Point: 1460°
Boiling Point: ca 2300°
Solubility: Practically insoluble in, but slowly hydrolyzed by water
Merck Reference: 14,1998
MDL No.: MFCD00010932
Formel:
CeF₃ Siedepunkt: ∼2300 °C Schmelzpunkt: 1460 °C Dichte: 6,16 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010932 CAS-Nummer: 7758-88-5 EINECS: 231-841-3 Merck Index: 14,1998 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...