4-Aminoantipyrin ≥98,0% analytisches Reagens
Lieferant: VWR Chemicals
21161.122EA
52.4
EUR
21161.122
21161.188
4-Aminoantipyrin ≥98,0% analytisches Reagens
4-Aminoantipyrin
Spezifikation Testergebnisse
Assay | Min. 98.0 % |
Suitable as phenol reagent | Passes test |
Melting point | 106 - 109 °C |
Ignition residue (SO4) | Max. 0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...