Brilliant black BN, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Food Black 1, Brilliant Black A, C.I. 28440, Black PN, Blue Black BN

211842-10G
211842-10GEA 96 EUR
211842-10G
Brilliant black BN, Sigma-Aldrich®
Brilliant black BN

Brilliant Black BN has been used: in micronucleus and comet assays to evaluate its genotoxic effect.as a quenching agent in calcium mobilization assay.

Formel: C₂₈H₁₇N₅O₁₄S₄
Molecular Weight: 867.66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078980
CAS-Nummer: 2519-30-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d37106ff581bf0ed78ffab4a31246b95e901533&vid=fdccaa9f128964781d2a99d27594514d44309901&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR