Calcimycin (A23187), Selectophore™, Funtion tested ionophore, Supelco®

Lieferant: Merck

Synonyme: Calcium Ionophore A23187, Calcium Ionophore III, Antibiotic A23187

MQ100

21186-25MG-F
21186-25MG-FEA 386 EUR
21186-25MG-F
Calcimycin (A23187), Selectophore™, Funtion tested ionophore, Supelco®
Calcimycin (A23187)

Calcium ionophore III (Calimycin;CA) is a divalent cation ionophore. It has ability to transport Ca2+ ions across membranes along a concentration gradient.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₂₉H₃₇N₃O₆
Molecular Weight: 523.63 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00151202
CAS-Nummer: 52665-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR