Ammoniaklösung 20%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

Synonyme: Ammoniumhydroxid, Ammoniak

21188.294EA 32.2 EUR
21188.294 21188.363
Ammoniaklösung 20%, AnalaR NORMAPUR® analytisches Reagens
Ammoniaklösung
Formel: NH₃
Dichte: 0,923 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066650
CAS-Nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Assay 18.0 - 22.0 %
Colouration Max. 10 APHA
Density (20/4) 0.910 - 0.930
Evaporation residue Max. 10 ppm
Silica Max. 10 ppm
Substances reducing KMnO4 (as O) Max. 4 ppm
Cl (Chloride) Max. 0.5 ppm
CO3 (Carbonate) Max. 10 ppm
PO4 (Phosphate) Max. 0.5 ppm
S (Sulphur) Max. 0.2 ppm
SO4 (Sulphate) Max. 20 ppm
Al (Aluminium) Max. 0.03 ppm
As (Arsenic) Max. 0.01 ppm
B (Boron) Max. 0.05 ppm
Ba (Barium) Max. 0.02 ppm
Be (Beryllium) Max. 0.02 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.5 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.3 ppm
Li (Lithium) Max. 0.01 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.01 ppm
Mo (Molybdenum) Max. 0.02 ppm
Na (Sodium) Max. 0.8 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.02 ppm
Sr (Strontium) Max. 0.01 ppm
Ti (Titanium) Max. 0.1 ppm
V (Vanadium) Max. 0.01 ppm
Zn (Zinc) Max. 1 ppm
Zr (Zirconium) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR