1,4-Bis(p-toluidino)-9,10-anthrachinon 95% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ200

211982-50G
211982-50GEA 131 EUR
211982-50G
1,4-Bis(p-toluidino)-9,10-anthrachinon 95% (Farbstoffgehalt), Sigma-Aldrich®
1,4-Bis(p-toluidino)-9,10-anthrachinon

Solvent Green 3, also known as Quinizarine Green SS (CI 61565) is an anthraquinone derivative. It is a green dye that imparts its colour to materials. Solvent Green 3 is made by condensing either leucoquinizarin or a mixture of leucoquinizarin and quinizarin with two moles of p-toluidine in the presence of boric acid.

Formel: C₂₈H₂₂N₂O₂
Schmelzpunkt: 220,..221 °C (Lit.)
MDL: MFCD00001195
CAS-Nummer: 128-80-3
EINECS: 204-909-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66d1c8743f8bb334b4674e4047b730c934354096&vid=66d1c8743f8bb334b4674e4047b730c934354096&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
C=0.01 G/L Acetone 624 - 644 nm
Extinction Coefficient ≥14000
Wavelength 394 - 414 nm
Extinction Coefficient, In Acetone ≥6000
Carbon 76.3 - 84.4 %
Nitrogen 6.3 - 7.0 %
TLC Consistent with Past Lots
Solubility (Turbidity) Turbid
Solubility (Color) c = 1%, Pyridine Dark Blue to Very Dark Blue and Dark, Blue-Green to Very Dark Blue-Green


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR