VWR®, Test Tubes without Rim, Thin Walled
Lieferant: VWR Collection
212-0744EA 106 EUR
212-0744 212-0745 212-0746 212-0747 212-0748 212-0749 212-0750 212-0751 212-0752
VWR®, Test Tubes without Rim, Thin Walled
Röhrchen Reagenzgläser- und Probenröhrchen
Test tubes made from borosilicate 7.1 glass, ideal for use as disposable.
  • Thin walled
  • Good chemical and physical properties
  • Not suitable for application related to strong heat or rapid temperature changes
An economically proven alternative in reasonable big packs.