Racks for cryovials and microtubes with lid
Lieferant: DELTALAB
G-050
212-8450EA 246 EUR
212-8450
Racks for cryovials and microtubes with lid
Gestelle Kühlgestelle
Styrofoam, white
  • Designed for long-term storage of microtubes and cryovials in the freezer
  • Alpha-numerically matrixed for better sample location
  • Stackable and lightweight