Microtube racks
Lieferant: DELTALAB
M-450
212-8940EA 30.5 EUR
212-8940
Microtube racks
Gestelle Reagenzglasgestelle
Racks, made of PP, designed for long term storage of samples at temperatures down to −90 °C.
  • With lift off telescopic lid
  • Lids with moulded alphanumeric identification system
  • Good chemical resistance
The four frosted side panels and a frosted rim on the edges of the lid provide additional labelling options.