Ammoniumacetat ≥98,0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens

Lieferant: VWR Chemicals
Achtung

21200.264EA 82.2 EUR
21200.264 21200.297 21200.366 21200.460
Ammoniumacetat ≥98,0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens
Ammoniumacetat
Formel: H₃CCOONH₄
Schmelzpunkt: 114 °C
Dichte: 1,17 g/cm³ (20 °C)
Flammpunkt: 136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013066
CAS-Nummer: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Min. 98.0 %
Appearance Colourless crystals
pH (25°C; 5 %) 6.7 - 7.3
Heavy metals (as Pb) Max. 2 ppm
Ignition residue (SO4) Max. 100 ppm
Insolubility in water Max. 50 ppm
Substances reducing KMnO4 (as HCOOH) Max. 50 ppm
Water Max. 2.0 %
Cl (Chloride) Max. 5 ppm
NO3 (Nitrate) Max. 10 ppm
SO4 (Sulphate) Max. 10 ppm
Ca (Calcium) Max. 10 ppm
Cd (Cadmium) Max. 0.5 ppm
Cu (Copper) Max. 0.5 ppm
Fe (Iron) Max. 2 ppm
Ni (Nickel) Max. 0.5 ppm
Pb (Lead) Max. 0.5 ppm
Zn (Zinc) Max. 0.5 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR