Calcium ionophore V ≥95%, Selectophore™ Ionophor, Supelco®

Lieferant: Merck

Synonyme: 10,19-Bis[(octadecylcarbamoyl)methoxyacetyl]-1,4,7,13,16-pentaoxa-10,19-diazacycloheneicosane, K23E1

M-Clarity™ Qualitätsstufe = MQ100

21203-25MG-F
21203-25MG-FEA 347 EUR
21203-25MG-F
Calcium ionophore V ≥95%, Selectophore™ Ionophor, Supelco®
Calcium ionophore V

Calcium ionophore V (K23E1) may be used in fabricating calcium on to the waveguide sensor to understand the selectivity of the specific ion and also measure its concentration.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₅₈H₁₁₂N₄O₁₁
Molecular Weight: 1041,53 g/mol
MDL: MFCD00282784
CAS-Nummer: 160563-01-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=628172c5fff48b985ce70a22471116b193a88b83&vid=b3267f28162f2c6102aabfd3fbc76f00c0ecac8e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR