Ammoniumhydrogencarbonat ≥97,5%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Achtung

Synonyme: Ammoniumbicarbonat

21219.292EA 135 EUR
21219.292
Ammoniumhydrogencarbonat ≥97,5%, AnalaR NORMAPUR® analytisches Reagens
Ammoniumhydrogencarbonat
Formel: NH₄HCO₃
Molecular Weight: 79,06 g/mol
Schmelzpunkt: 35 °C
Dichte: 1,57 g/cm³ (20 °C)
MDL: MFCD00012138
CAS-Nummer: 1066-33-7
EINECS: 213-911-5
Merck Index: 13,00497

Bestellen

Spezifikation Testergebnisse

Assay Min. 97.5 %
Heavy metals (as Pb) Max. 5 ppm
Ignition residue (SO4) Max. 50 ppm
Insolubility in water Max. 50 ppm
Cl (Chloride) Max. 5 ppm
PO4 (Phosphate) Max. 10 ppm
SO4 (Sulphate) Max. 20 ppm
Fe (Iron) Max. 2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR