Ammoniumcer(IV)sulfat Dihydrat ≥94,0%, AnalaR NORMAPUR® analytisches Reagens
Lieferant: VWR Chemicals
Synonyme:
Cer(IV)ammoniumsulfat Dihydrat, tetra-Ammoniumcertetrakis(sulfat) Dihydrat
21232.151EA
90.4
EUR
21232.151
Ammoniumcer(IV)sulfat Dihydrat ≥94,0%, AnalaR NORMAPUR® analytisches Reagens
Ammoniumcer(IV)sulfat Dihydrat
Formel:
(NH₄)₂Ce(SO₄)₄·2H₂O Molecular Weight: 632.55 g/mol Schmelzpunkt: 130 |
MDL:
MFCD00149417 CAS-Nummer: 10378-47-9 EINECS: 231-567-4 |
Spezifikation Testergebnisse
Assay | Min. 94.0 % |
Heavy metals (as Pb) | Max. 50 ppm |
Insolubility in diluted H2SO4 | Max. 50 ppm |
Cl (Chloride) | Max. 10 ppm |
PO4 (Phosphate) | Max. 50 ppm |
Fe (Iron) | Max. 50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...