Direct Red 23, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

212490-50G
212490-50GEA 134 EUR
212490-50G
Direct Red 23, Sigma-Aldrich®
Direct Red 23

Direct Red 23 has been used to stain water-hydrated seeds.

Formel: C₃₅H₂₅N₇Na₂OS₂
Molecular Weight: 813,72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003946
CAS-Nummer: 3441-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d62ab5e9f72469ef7b3ff9cfa9f00b204433d37b&vid=7d6a7e72d69d47cf209c8986efe520e531a5a970&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR