Ammoniumdihydrogenphosphat ≥99,0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Lieferant: VWR Chemicals
21305.290EA
81.1
EUR
21305.290
21305.260
Ammoniumdihydrogenphosphat ≥99,0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Ammoniumdihydrogenphosphat
Formel:
NH₄H₂PO₄ Schmelzpunkt: 190 °C Dichte: 1,8 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003396 CAS-Nummer: 7722-76-1 EINECS: 231-764-5 REACH: 01-2119488166-29 Merck Index: 13,00547 |
Spezifikation Testergebnisse
Assay | Min. 99.0 % |
pH (2,3 %) | About 4.2 |
pH (25°C; 5 %) | 3.8 - 4.4 |
Heavy metals (as Pb) | Max. 5 ppm |
Insolubility in water | Max. 50 ppm |
Cl (Chloride) | Max. 5 ppm |
NO3 (Nitrate) | Max. 10 ppm |
SO4 (Sulphate) | Max. 50 ppm |
Ca (Calcium) | Max. 10 ppm |
Fe (Iron) | Max. 10 ppm |
K (Potassium) | Max. 50 ppm |
Mg (Magnesium) | Max. 5 ppm |
Na (Sodium) | Max. 50 ppm |
Conforms to ACS | Passes test |
Conforms to Reag. Ph.Eur. | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...