di-Ammoniumhydrogenphosphat ≥98%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Chemicals

21311.234EA 38.5 EUR
21311.234 21311.291 21311.460
di-Ammoniumhydrogenphosphat ≥98%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
di-Ammoniumhydrogenphosphat
Formel: (NH₄)₂HPO₄
Schmelzpunkt: 185 °C
Dichte: 1,619 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010891
CAS-Nummer: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546

Bestellen

Spezifikation Testergebnisse

Assay Min. 98 %
pH (20°C) About 8
pH (25°C; 5 %) 7.7 - 8.1
Heavy metals (as Pb) Max. 0.001 %
Insolubility in water Max. 50 ppm
Cl (Chloride) Max. 0.001 %
SO4 (Sulphate) Max. 0.01 %
NO3 (Nitrate) Max. 0.003 %
Ca (Calcium) Max. 10 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 0.005 %
Mg (Magnesium) Max. 5 ppm
Na (Sodium) Max. 0.005 %
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR