Messkolben, Klasse B, Azlon®
Lieferant: SciLabware
FDF018N FDF024N FDF012N FDF030N FDF036N
214-0116EA 22.1 EUR
214-0116 214-0117 214-0115 214-0118 214-0247
Messkolben, Klasse B, Azlon®
Kolben Messkolben
PP, durchscheinend, mit PE-Stopfen
  • Class B tolerances with a clear easy to read calibration line
  • Feature a mixing chamber at the top of each flask's neck
  • Exposure to temperatures above 60 °C may affect accuracy
  • Cleaning with mild alkaline detergents recommended up to 60 °C
Standards: ISO 1042