Volumetric flasks, class B, red markings
Lieferant: VITLAB
676895
214-0227EA 23.9 EUR
214-0227
Volumetric flasks, class B, red markings
Kolben Messkolben
PMP, crystal clear, with screw cap.
  • With ring mark individually calibrated to 'In'
  • Thermal stress up to 121 °C (autoclaving) does not permanently exceed the tolerance limit
  • To preserve markings. cleaning at no higher than 60 °C is recommended
Standards: Class B tolerances according to DIN EN ISO 1042