Ring weights for flasks and vessels
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
W-1252
214-1917EA 96.8 EUR
214-1917
Ring weights for flasks and vessels
Gewichte Beschwerungsgewichte
Open C-shaped flask weight ring with thick PVC coated. Floating can be avoided by placing this ring around the neck of the vessels.
  • High chemical and thermal resistance
  • No floating
  • Falling over is impossible