Holder for Flask immersion weights, ring type, Poxygrid®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F183071000
214-1918EA 75.3 EUR
214-1918
Holder for Flask immersion weights, ring type, Poxygrid®, SP Bel-Art
Gewichte Beschwerungsgewichte
This wire formed, epoxy coated holder keeps lead ring weights organised and handy.
  • Small footprint
  • Base Ø: 190 mm
Larger stack of 171 mm height is neatly designed on tapered centre.