Glycerin (pflanzliche Basis) 99,0-101,0% (durch Trockensubstanz) USP, J.T.Baker®

Lieferant: Avantor

Synonyme: 1,2,3-Propantriol, Glycerol

2142-01EA 142.48 EUR
2142-01 2142-03 2142-07 2142-09
Glycerin (pflanzliche Basis) 99,0-101,0% (durch Trockensubstanz) USP, J.T.Baker®
Glycerin

  • GMP manufactured product

Formel: HOCH₂CH(OH)CH₂OH
Siedepunkt: 290 °C (1013 hPa)
Schmelzpunkt: 18,6 °C
Dichte: 1,26 g/cm³ (20 °C)
Flammpunkt: 177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004722
CAS-Nummer: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 99.0 - 101.0%
Meets U.S.P Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging ≤0.10 %
Bulk Pharmaceutical Chemical ≤0.10 %
USP - Assay (as HOCH₂CHOHCH₂OH) (calculated on anhydrous basis) 99.0 - 101.0 %
USP - Identification A Passes Test
USP - Identification B - Diethylene Glycol ≤0.10 %
USP - Identification B - Ethylene Glycol ≤0.10 %
USP - Identification C Passes Test
USP - Specific Gravity at 25°/25°C ≥1.249
USP - Color Passes Test
USP - Residue on Ignition ≤0.01 %
USP - Chloride (Cl) ≤0.001 %
USP - Sulfate (SO₄) ≤0.002 %
USP - Chlorinated Compounds (as Cl) ≤0.003 %
USP - Fatty Acids and Esters Passes Test
USP - Water (H₂O) ≤5.0 %
USP - Related Compounds: Individual Impurity ≤0.1 %
USP - Related Compo unds: Total Impurities ≤1.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR