Kieselgel 40 6-14 mesh, technical, Supelco®

Lieferant: Merck

214426-1KG
214426-1KGEA 62.6 EUR
214426-1KG
Kieselgel 40 6-14 mesh, technical, Supelco®
Silica gel
Formel: SiO₂
Molecular Weight: 60.08 g/mol
Siedepunkt: 2230 °C
Schmelzpunkt: > 1600 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=402027dbf89b187bb319318cfea9e820b4a1ef77&vid=09421e7fa7d948e6820204b8322caaf3b6ebf98d&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR