Kieselgel, Davisil® Grade 923 pore size 30 Å, 100-200 mesh, hochrein, Supelco®

Lieferant: Merck

214477-250G 214477-50G
214477-250GEA 239 EUR
214477-250G 214477-50G
Kieselgel, Davisil® Grade 923 pore size 30 Å, 100-200 mesh, hochrein, Supelco®
Silica gel
Formel: SiO₂
Siedepunkt: 2230 °C
Schmelzpunkt: > 1600 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff484c390424ecbc604290464e00fc361ea19178&vid=ab04e2801dffc54a5f5c0faae685b6bb0ed3cf74&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR