Buckets, Round
Lieferant: SEMADENI
0699
215-0274EA 44.8 EUR
215-0274
Buckets, Round
Eimer
Round buckets, made of HDPE, natural, with graduation, handle and spout.
  • Sturdy
  • Good chemical resistance
  • Hot water resistant