Weithalsflaschen mit Schraubverschluss, EP-Tox, Nalgene®
Lieferant: Thermo Fisher Scientific
2101-2200
215-1031EA 877 EUR
215-1031
Weithalsflaschen mit Schraubverschluss, EP-Tox, Nalgene®
Flaschen Kunststoffflaschen
FEP, translucent with white PP screw cap with PFA liner.
  • Maximum chemical resistance
  • Temperature resistant from −105 to +150 °C, autoclavable
  • Leakproof closure
Specifically designed for EPA method 1311 TCLP Toxicity Characteristic Leaching Procedure. Excellent chemical and temperature resistance for large-sized samples. Very durable bottles for low and high-temperature applications, for trace analysis and organic solvents.