Spritzflaschen, Enghals
Lieferant: KARTELL
179 180 182 183 185
215-1355EA 6 EUR
215-1355 215-1356 215-1357 215-6413 215-6414
Spritzflaschen, Enghals
Flaschen Spritzflaschen
PE, translucent.
  • Flexible spout
  • Removable dispensing jet
  • Good chemical resistance
Narrow mouth wash bottles with screw cap.