VWR®, Nährmedienflaschen mit Schraubverschluss
Lieferant: VWR Collection
215-2203EA 145 EUR
215-2203 215-2204 215-2205 215-2206 215-2210 215-2211 215-2924
VWR®, Nährmedienflaschen mit Schraubverschluss
Flaschen Zellkulturflaschen
PC, autoclavable, narrow mouth bottles with standard PP caps.
  • Ideal for steam sterilisation
  • Safe and shatterproof
  • Chemically inert
VWR® media bottles are an ideal replacement for all borosilicate glass bottles. They are specifically designed for every laboratory application requiring terminal sterilisation (autoclaving), storage, transportation and production of laboratory biological fluids. These bottles are available in square and round shapes to accommodate various packaging needs.
The closure system is to be guaranteed leakproof, and theMore Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable