Bottles, wide neck, HDPE, round, with screw cap, lab quality, Nalgene®
Lieferant: Thermo Fisher Scientific
312104-0008 312104-0004 312104-0002 312104-0001 312104-0016 312104-0032
215-2768EA 635 EUR
215-2768 215-2767 215-2766 215-2574 215-2769 215-2770
Bottles, wide neck, HDPE, round, with screw cap, lab quality, Nalgene®
Flaschen Kunststoffflaschen
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Leakproof closure
Can be used anywhere in the laboratory and for sample collection. Wide opening allows rapid filling and dispensing of powders, pastes and liquids.
Standards: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Verpackung: Bottles and closures are packed in separate polybags within the same carton.