Pouring rings
Lieferant: SciLabware
4516/32D 4516/45D 4516/80D
BIBB4516/32DEA 10.3 EUR
BIBB4516/32D 215-3138 BIBB4516/80D
Pouring rings
Verschlüsse Flaschenverschlüsse
Pouring rings, made of Polypropylene, for laboratory/media bottles.
  • Ensures drip-free pouring
  • Very good chemical resistance
  • Autoclavable up to 140 °C