Jars, wide mouth, with screw cap, Corning® Gosselin™
Lieferant: Corning
PR1000-02 PR1050-01 PR500-02 PR800-01
215-3242EA 550 EUR
215-3242 215-3243 215-3244 GOSSPR800-01
Jars, wide mouth, with screw cap, Corning® Gosselin™
Dosen Dosen, Universal
PP, white.
  • Straight
  • With screw cap
  • Food contact approved
These round jars are ideal for use in sampling applications in clinical and industrial environments.