Spray bottles
Lieferant: BURKLE
0309-9900
215-4364EA 0 EUR
215-4364
Spray bottles
Flaschen Sprühflaschen
Basic spray bottles are suitable for occasional spraying in various industries.
  • Spray mist is ideal for spraying various liquids over a large area
  • Pressureless spray bottle is made of HDPE
  • Nozzle functions: Stop and spray
Bottle components in contact with media include PP, PE, TPE, and POM.