Fuchsin basisch in ACS analysereinem Wasser ≥85% (Farbstoffgehalt), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
Basic fuchsin is a triaminotriphenylmethane dye that is widely used to stain muscle, mitochondria, and collagen. It is generally a mixture of pararosaniline (C.I. 42500), rosaniline (C.I. 42510), magenta II and new fuchsine (CI 42520).
- Acid-fast staining
- Periodic Acid-Schiff
- Feulgen stain for DNA
- ENDO culture medium
- Leifson's method for bacterial flagella
Basic fuchsin displays anesthetic, bactericidal (gram-positive), and fungicidal properties.
Formel:
C₂₀H₂₀ClN₃ Schmelzpunkt: 235 °C Dichte: 1,22 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012569 CAS-Nummer: 632-99-5 EINECS: 211-189-6 Merck Index: 12,05686 |
|
Spezifikation Testergebnisse
Appearance (Colour) | Green |
Appearance (Form) | Powder or Crystals |
Infrared spectrum | Conforms to Structure |
Wavelength c = 0.003 g/L in 50% Ethanol |
543 - 549 nm |
Extinction Coefficient | ≥17000 |
Wavelength | 286 - 292 nm |
Carbon | 59.9 - 81.0% |
Nitrogen | 11.0 - 14.9% |
Purity (HPLC) | ≥85.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...