1,4,5,8-Tetraamino-9,10-anthrachinon 30% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Disperse Blue 1

215643-5G
215643-5GEA 93.2 EUR
215643-5G
1,4,5,8-Tetraamino-9,10-anthrachinon 30% (Farbstoffgehalt), Sigma-Aldrich®
1,4,5,8-Tetraamino-9,10-anthrachinon

Disperse blue 1 is a grey to black powder. Disperse dyes are known to be insoluble in water and are applied to the substrate as suspensions or dispersions by dissolving in hydrophobic regions of the polymers making up the fiber.

Formel: C₁₄H₁₂N₄O₂
Molecular Weight: 268,27 g/mol
Schmelzpunkt: 332 °C
Lagertemperatur: Room Temperature
MDL: MFCD00001225
CAS-Nummer: 2475-45-8
EINECS: 219-603-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0307f0a528bce74de2d373ece5fbaeaae126a4a4&vid=0307f0a528bce74de2d373ece5fbaeaae126a4a4&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Grey to black powder
Infrared spectrum Conforms to structure
UV-visible spectrum 0.01 g/L in 50% ethanol w/1 drop 1N NaOH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR