di-Phosphorpentoxid 99+% zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

215751000 215755000 215750025
215751000.EA 21.4 EUR
215751000. 215755000. 215750025.
di-Phosphorpentoxid 99+% zur Analyse
di-Phosphorpentoxid
Formel: P₂O₅
Schmelzpunkt: 340…360 °C (Subl.)
Dichte: 2,253 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011440
CAS-Nummer: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8,II
Merck Index: 13,07441

Bestellen

Spezifikation Testergebnisse

Appearance White Deliquescent powder
Titration with NaOH ≥99.0 %
Chloride (Cl) ≤0.001 %
Nitrate (NO3) ≤0.0005 %
Ammonium (NH4) ≤0.001 %
Arsenic (As) ≤0.01 %
Cadmium (Cd) ≤0.0005 %
Calcium (Ca) ≤0.005 %
Cobalt (Co) ≤0.0005 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.001 %
Lead (Pb) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Manganese (Mn) ≤0.0005 %
Nickel (Ni) ≤0.0005 %
Potassium (K) ≤0.002 %
Sodium (Na) ≤0.005 %
Zinc (Zn) ≤0.0005 %
Heavy metals (as Pb) ≤0.005 %
Reducing substance (P2O3) ≤0.02 %
Total nitrogen (as N) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR