Ballonflaschen, rechteckig, mit Hahn, Nalgene®
Lieferant: Thermo Fisher Scientific
DS2327-0020 DS2327-0050
216-1222EA 262 EUR
216-1222 216-1223
Ballonflaschen, rechteckig, mit Hahn, Nalgene®
Flaschen Ballons
Made of FLPE with TFE spigot and fluorinated PP screw cap.
  • Graduated, tough rigid and translucent
  • Space-saving rectangular shape
  • Attached stainless steel handle
Excellent for storing and dispensing a variety of solvents; useful for collecting and dispensing distilled water. Fluorinated surface (inside and outside) improves barrier properties; reduces solvent absorption and permeation.