Ice pans and buckets, multi-purpose, Corning®
Lieferant: Corning
432093 432094 432095 432096 432097 432098 432099 432100 432101 432102 432103 432104 432105 432106 432107 432108 432109 432110 432111 432112 432113 432114 432115 432116 432117 432118 432119 432120 432121 432122 432123 432124 432125 432126 432127 432128 432129 432130 432131 432132 432133 432134 432135
479-1977EA 196 EUR
479-1977 479-1978 479-1979 479-1980 479-1981 479-1982 479-1983 479-1984 479-1985 479-1986 216-1941 216-1942 216-1943 216-1944 216-1945 216-1946 216-1947 479-1987 479-1988 479-1989 479-1990 479-1991 479-1992 479-1993 479-1994 479-1995 479-1996 479-1997 479-1998 216-1948 216-1949 216-1950 216-1951 216-1952 216-1953 216-1954 479-1999 479-2013 479-2014 479-2015 479-2016 479-2017 479-2018
Ice pans and buckets, multi-purpose, Corning®
Eisbehälter
Ethylene vinyl acetate (EVA) foam.
  • Solid, one piece moulded, unbreakable
  • Resistant to highly acidic and basic solutions, salts, alcohol, acetone, chloroform, phenol, etc
  • Insulative properties and resistant to temperatures from −196 to 93 °C
Ideal for use with ice, dry ice, liquid nitrogen, saline solution and acetone.
Corning ice pans and buckets are multi-purpose pans and buckets that are a great non-toxic alternative to conventional ice pans.
EVA has little or no odor, is inert and used in biomedical engineering applications such as drugMore Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable Unbedenklicher für Mensch und Umwelt